2,4-Dimethylbenzenesulfonohydrazide

Supplier: Matrix Scientific

029922-500MG
101829-120EA 193.85 USD
101829-120
2,4-Dimethylbenzenesulfonohydrazide
2,4-Dimethylbenzenesulfonohydrazide

Hazard: Irritant

Formula: C₈H₁₂N₂O₂S
MDL Number: MFCD03423366
CAS Number: 18626-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR