2,5-Dimethylbenzenesulfonohydrazide

Supplier: Matrix Scientific

029921-500MG
101829-118EA 193.85 USD
101829-118
2,5-Dimethylbenzenesulfonohydrazide
2,5-Dimethylbenzenesulfonohydrazide

Hazard: Irritant

Formula: C₈H₁₂N₂O₂S
MDL Number: MFCD02710371
CAS Number: 38045-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR