4-tert-Butylbenzenesulfonohydrazide

Supplier: Matrix Scientific

029917-500MG
101829-110EA 193.85 USD
101829-110
4-tert-Butylbenzenesulfonohydrazide
4-tert-Butylbenzenesulfonohydrazide

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₂S
MDL Number: MFCD00229050
CAS Number: 65604-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR