4-Butylbenzenesulfonohydrazide

Supplier: Matrix Scientific

029916-500MG
101829-108EA 144.9 USD
101829-108
4-Butylbenzenesulfonohydrazide
4-Butylbenzenesulfonohydrazide

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₂S
MDL Number: MFCD03423362
CAS Number: 774575-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR