4-Isopropylbenzenesulfonohydrazide
Supplier: Matrix Scientific
029915-500MG
101829-106EA
193.85
USD
101829-106
4-Isopropylbenzenesulfonohydrazide
4-Isopropylbenzenesulfonohydrazide
Hazard: Irritant
Formula:
C₉H₁₄N₂O₂S |
MDL Number:
MFCD01004370 CAS Number: 219741-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...