4-Ethylbenzenesulfonohydrazide

Supplier: Matrix Scientific

029913-500MG
101829-102EA 144.9 USD
101829-102
4-Ethylbenzenesulfonohydrazide
4-Ethylbenzenesulfonohydrazide

Hazard: Irritant

Formula: C₈H₁₂N₂O₂S
MDL Number: MFCD01231410
CAS Number: 18684-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR