4-Ethoxybenzenesulfonohydrazide

Supplier: Matrix Scientific

029912-500MG
101829-100EA 193.85 USD
101829-100
4-Ethoxybenzenesulfonohydrazide
4-Ethoxybenzenesulfonohydrazide

Hazard: Irritant

Formula: C₈H₁₂N₂O₃S
MDL Number: MFCD00519218
CAS Number: 24924-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR