N-[4-(Hydrazinosulfonyl)phenyl]acetamide

Supplier: Matrix Scientific

029911-500MG
101829-098EA 144.9 USD
101829-098
N-[4-(Hydrazinosulfonyl)phenyl]acetamide
N-[4-(Hydrazinosulfonyl)phenyl]acetamide

Hazard: Irritant

Formula: C₈H₁₁N₃O₃S
MDL Number: MFCD00025103
CAS Number: 3989-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR