2-(2-Fluorophenoxy)acetohydrazide
Supplier: Matrix Scientific
029906-500MG
101829-088EA
146.15
USD
101829-088
2-(2-Fluorophenoxy)acetohydrazide
2-(2-Fluorophenoxy)acetohydrazide
Hazard: Irritant
Formula:
C₈H₉FN₂O₂ |
MDL Number:
MFCD02295759 CAS Number: 380426-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...