2-Hydroxy-2-methylpropanohydrazide

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-2-methylpropanehydrazide

029905-500MG
101829-086EA 193.85 USD
101829-086
2-Hydroxy-2-methylpropanohydrazide
2-Hydroxy-2-methylpropanohydrazide

Hazard: Irritant

Formula: C₄H₁₀N₂O₂
MW: 118.14 g/mol
MDL Number: MFCD00031406
CAS Number: 42596-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR