3-Hydroxybutanohydrazide

Supplier: Matrix Scientific

029904-500MG
101829-084EA 95.38 USD
101829-084
3-Hydroxybutanohydrazide
3-Hydroxybutanohydrazide

Hazard: Irritant

Formula: C₄H₁₀N₂O₂
MDL Number: MFCD00087370
CAS Number: 24534-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR