5-[(4-Chloro-3,5-dimethylphenoxy)methyl]-2-furohydrazide

Supplier: Matrix Scientific

029903-500MG
101829-082EA 146.15 USD
101829-082
5-[(4-Chloro-3,5-dimethylphenoxy)methyl]-2-furohydrazide
5-[(4-Chloro-3,5-dimethylphenoxy)methyl]-2-furohydrazide

Hazard: Irritant

Formula: C₁₄H₁₅ClN₂O₃
MDL Number: MFCD05861693
CAS Number: 861508-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR