2-Ethylhexanohydrazide

Supplier: Matrix Scientific

029902-500MG
101829-080EA 146.15 USD
101829-080
2-Ethylhexanohydrazide
2-Ethylhexanohydrazide

Hazard: Irritant

Formula: C₈H₁₈N₂O
MDL Number: MFCD03014512
CAS Number: 41632-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR