2-(1,3-Benzothiazol-2-ylthio)propanohydrazide

Supplier: Matrix Scientific

029901-500MG
101829-078EA 193.85 USD
101829-078
2-(1,3-Benzothiazol-2-ylthio)propanohydrazide
2-(1,3-Benzothiazol-2-ylthio)propanohydrazide

Hazard: Irritant

Formula: C₁₀H₁₁N₃OS₂
MDL Number: MFCD02161583
CAS Number: 99055-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR