2-(2,4-Dibromophenoxy)propanohydrazide

Supplier: Matrix Scientific

029899-500MG
101829-074EA 146.15 USD
101829-074
2-(2,4-Dibromophenoxy)propanohydrazide
2-(2,4-Dibromophenoxy)propanohydrazide

Hazard: Irritant

Formula: C₉H₁₀Br₂N₂O₂
MDL Number: MFCD03423275
CAS Number: 862705-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR