2-Methylpropanohydrazide

Supplier: Matrix Scientific

Synonyms: IsobutyrohydrazideIsobutyric acid Hydrazide

029895-5G
101829-070EA 69.23 USD
101829-070
2-Methylpropanohydrazide
2-Methylpropanohydrazide

Hazard: Irritant

Formula: C₄H₁₀N₂O
MW: 102.14 g/mol
Melting Pt: 98…100 °C
MDL Number: MFCD00025122
CAS Number: 3619-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR