4-Chloro-1,3-dimethyl-1H-pyrazole-5-carbohydrazide

Supplier: Matrix Scientific

029892-500MG
101829-064EA 290.77 USD
101829-064
4-Chloro-1,3-dimethyl-1H-pyrazole-5-carbohydrazide
4-Chloro-1,3-dimethyl-1H-pyrazole-5-carbohydrazide

Hazard: Irritant

Formula: C₆H₉ClN₄O
MDL Number: MFCD02090990
CAS Number: 676348-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR