4-Bromo-1-methyl-1H-pyrazole-3-carbohydrazide

Supplier: Matrix Scientific

029891-500MG
101829-062EA 290.77 USD
101829-062
4-Bromo-1-methyl-1H-pyrazole-3-carbohydrazide
4-Bromo-1-methyl-1H-pyrazole-3-carbohydrazide

Hazard: Irritant

Formula: C₅H₇BrN₄O
MDL Number: MFCD02253808
CAS Number: 400878-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR