4-Chloro-1-methyl-1H-pyrazole-5-carbohydrazide

Supplier: Matrix Scientific

029890-500MG
101829-060EA 217.35 USD
101829-060
4-Chloro-1-methyl-1H-pyrazole-5-carbohydrazide
4-Chloro-1-methyl-1H-pyrazole-5-carbohydrazide

Hazard: Irritant

Formula: C₅H₇ClN₄O
MDL Number: MFCD01459852
CAS Number: 400878-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR