1-Ethyl-1H-pyrazole-5-carbohydrazide

Supplier: Matrix Scientific

029887-500MG
101829-054EA 217.35 USD
101829-054
1-Ethyl-1H-pyrazole-5-carbohydrazide
1-Ethyl-1H-pyrazole-5-carbohydrazide

Hazard: Irritant

Formula: C₆H₁₀N₄O
MDL Number: MFCD03423248
CAS Number: 1001755-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR