1-Ethyl-1H-pyrazole-3-carbohydrazide

Supplier: Matrix Scientific

029886-500MG
101829-052EA 290.77 USD
101829-052
1-Ethyl-1H-pyrazole-3-carbohydrazide
1-Ethyl-1H-pyrazole-3-carbohydrazide

Hazard: Irritant

Formula: C₆H₁₀N₄O
MDL Number: MFCD03423247
CAS Number: 1006334-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR