4-Iodo-1-methyl-1H-pyrazole-3-carbohydrazide

Supplier: Matrix Scientific

029885-500MG
101829-050EA 290.77 USD
101829-050
4-Iodo-1-methyl-1H-pyrazole-3-carbohydrazide
4-Iodo-1-methyl-1H-pyrazole-3-carbohydrazide

Hazard: Irritant

Formula: C₅H₇IN₄O
MDL Number: MFCD00466328
CAS Number: 1001822-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR