2-(2,4,6-Trichlorophenoxy)acetohydrazide

Supplier: Matrix Scientific

029883-500MG
101829-046EA 221.54 USD
101829-046
2-(2,4,6-Trichlorophenoxy)acetohydrazide
2-(2,4,6-Trichlorophenoxy)acetohydrazide

Hazard: Irritant

Formula: C₈H₇Cl₃N₂O₂
MDL Number: MFCD03423246
CAS Number: 190588-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR