2-(4-Methylphenyl)cyclopropanecarbohydrazide

Supplier: Matrix Scientific

029881-500MG
101829-042EA 290.77 USD
101829-042
2-(4-Methylphenyl)cyclopropanecarbohydrazide
2-(4-Methylphenyl)cyclopropanecarbohydrazide

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
MDL Number: MFCD02253832
CAS Number: 438219-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR