2-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)acetohydrazide

Supplier: Matrix Scientific

029880-500MG
101829-040EA 290.77 USD
101829-040
2-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)acetohydrazide
2-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)acetohydrazide

Hazard: Irritant

Formula: C₇H₁₁BrN₄O
MW: 247.1 g/mol
MDL Number: MFCD00173770
CAS Number: 175137-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR