5-[(4-Bromo-3-nitro-1H-pyrazol-1-yl)methyl]-2-furohydrazide

Supplier: Matrix Scientific

029879-500MG
101829-038EA 217.35 USD
101829-038
5-[(4-Bromo-3-nitro-1H-pyrazol-1-yl)methyl]-2-furohydrazide
5-[(4-Bromo-3-nitro-1H-pyrazol-1-yl)methyl]-2-furohydrazide

Hazard: Irritant

Formula: C₉H₈BrN₅O₄
MDL Number: MFCD02253821
CAS Number: 512810-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR