5-(1H-Pyrazol-1-ylmethyl)-2-furohydrazide

Supplier: Matrix Scientific

029878-500MG
101829-036EA 217.35 USD
101829-036
5-(1H-Pyrazol-1-ylmethyl)-2-furohydrazide
5-(1H-Pyrazol-1-ylmethyl)-2-furohydrazide

Hazard: Irritant

Formula: C₉H₁₀N₄O₂
MDL Number: MFCD02253818
CAS Number: 457051-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR