5-[(2-Isopropyl-5-methylphenoxy)methyl]-2-furohydrazide

Supplier: Matrix Scientific

029874-500MG
101829-028EA 290.77 USD
101829-028
5-[(2-Isopropyl-5-methylphenoxy)methyl]-2-furohydrazide
5-[(2-Isopropyl-5-methylphenoxy)methyl]-2-furohydrazide

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₃
MDL Number: MFCD02090997
CAS Number: 438222-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR