5-[(4-Propylphenoxy)methyl]-2-furohydrazide

Supplier: Matrix Scientific

029873-500MG
101829-026EA 217.35 USD
101829-026
5-[(4-Propylphenoxy)methyl]-2-furohydrazide
5-[(4-Propylphenoxy)methyl]-2-furohydrazide

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₃
MDL Number: MFCD02090996
CAS Number: 364743-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR