5-[(2,5-Dichlorophenoxy)methyl]-2-furohydrazide

Supplier: Matrix Scientific

029872-500MG
101829-024EA 290.77 USD
101829-024
5-[(2,5-Dichlorophenoxy)methyl]-2-furohydrazide
5-[(2,5-Dichlorophenoxy)methyl]-2-furohydrazide

Hazard: Irritant

Formula: C₁₂H₁₀Cl₂N₂O₃
MDL Number: MFCD02055010
CAS Number: 406470-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR