2-(4-Methoxyphenyl)-6,8-dimethylquinoline-4-carbohydrazide

Supplier: Matrix Scientific

029868-500MG
101829-016EA 217.35 USD
101829-016
2-(4-Methoxyphenyl)-6,8-dimethylquinoline-4-carbohydrazide
2-(4-Methoxyphenyl)-6,8-dimethylquinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₉H₁₉N₃O₂
MDL Number: MFCD03420517
CAS Number: 590360-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR