2,8-Dimethylquinoline-4-carbohydrazide

Supplier: Matrix Scientific

029867-500MG
101829-014EA 290.77 USD
101829-014
2,8-Dimethylquinoline-4-carbohydrazide
2,8-Dimethylquinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₂H₁₃N₃O
MDL Number: MFCD03420481
CAS Number: 590376-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR