2-(4-tert-Butylphenyl)-8-methylquinoline-4-carbohydrazide

Supplier: Matrix Scientific

029865-500MG
101829-010EA 290.77 USD
101829-010
2-(4-tert-Butylphenyl)-8-methylquinoline-4-carbohydrazide
2-(4-tert-Butylphenyl)-8-methylquinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₂₁H₂₃N₃O
MDL Number: MFCD03420454
CAS Number: 590376-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR