2-Cyclopropyl-6-methylquinoline-4-carbohydrazide

Supplier: Matrix Scientific

029864-500MG
101829-008EA 290.77 USD
101829-008
2-Cyclopropyl-6-methylquinoline-4-carbohydrazide
2-Cyclopropyl-6-methylquinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₄H₁₅N₃O
MDL Number: MFCD03420432
CAS Number: 956576-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR