2-(4-Isopropoxyphenyl)-6-methylquinoline-4-carbohydrazide

Supplier: Matrix Scientific

029862-500MG
101829-004EA 290.77 USD
101829-004
2-(4-Isopropoxyphenyl)-6-methylquinoline-4-carbohydrazide
2-(4-Isopropoxyphenyl)-6-methylquinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₂₀H₂₁N₃O₂
MDL Number: MFCD03420417
CAS Number: 590355-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR