2-(4-Isopropylphenyl)-6-methylquinoline-4-carbohydrazide
Supplier: Matrix Scientific
029858-500MG
101828-996EA
217.35
USD
101828-996
2-(4-Isopropylphenyl)-6-methylquinoline-4-carbohydrazide
2-(4-Isopropylphenyl)-6-methylquinoline-4-carbohydrazide
Hazard: Irritant
Formula:
C₂₀H₂₁N₃O |
MDL Number:
MFCD03420397 CAS Number: 438221-46-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...