2-(4-Ethylphenyl)-6-methylquinoline-4-carbohydrazide

Supplier: Matrix Scientific

029857-500MG
101828-994EA 290.77 USD
101828-994
2-(4-Ethylphenyl)-6-methylquinoline-4-carbohydrazide
2-(4-Ethylphenyl)-6-methylquinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₉H₁₉N₃O
MDL Number: MFCD03420395
CAS Number: 588715-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR