6-Methyl-2-(4-methylphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029856-500MG
101828-992EA 217.35 USD
101828-992
6-Methyl-2-(4-methylphenyl)quinoline-4-carbohydrazide
6-Methyl-2-(4-methylphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₈H₁₇N₃O
MDL Number: MFCD03420391
CAS Number: 588696-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR