6-Bromo-2-(4-isopropylphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029855-500MG
101828-990EA 290.77 USD
101828-990
6-Bromo-2-(4-isopropylphenyl)quinoline-4-carbohydrazide
6-Bromo-2-(4-isopropylphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₉H₁₈BrN₃O
MDL Number: MFCD03420755
CAS Number: 956576-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR