2-(3,4-Diethoxyphenyl)acetohydrazide

Supplier: Matrix Scientific

029853-500MG
101828-986EA 109.25 USD
101828-986
2-(3,4-Diethoxyphenyl)acetohydrazide
2-(3,4-Diethoxyphenyl)acetohydrazide

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₃
MDL Number: MFCD03419414
CAS Number: 91908-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR