2-(2-Chloro-6-fluorophenyl)acetohydrazide

Supplier: Matrix Scientific

029851-500MG
101828-982EA 146.15 USD
101828-982
2-(2-Chloro-6-fluorophenyl)acetohydrazide
2-(2-Chloro-6-fluorophenyl)acetohydrazide

Hazard: Irritant

Formula: C₈H₈ClFN₂O
MDL Number: MFCD03423204
CAS Number: 669740-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR