3-Chloro-4-fluoro-1-benzothiophene-2-carbohydrazide

Supplier: Matrix Scientific

029849-500MG
101828-978EA 290.77 USD
101828-978
3-Chloro-4-fluoro-1-benzothiophene-2-carbohydrazide
3-Chloro-4-fluoro-1-benzothiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₉H₆ClFN₂OS
MDL Number: MFCD03423202
CAS Number: 774586-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR