3-Methyl-1-benzofuran-2-carbohydrazide

Supplier: Matrix Scientific

029848-500MG
101828-976EA 290.77 USD
101828-976
3-Methyl-1-benzofuran-2-carbohydrazide
3-Methyl-1-benzofuran-2-carbohydrazide

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD03109883
CAS Number: 53524-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR