5-Ethylthiophene-2-carbohydrazide

Supplier: Matrix Scientific

029847-500MG
101828-974EA 290.77 USD
101828-974
5-Ethylthiophene-2-carbohydrazide
5-Ethylthiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₇H₁₀N₂OS
MDL Number: MFCD01627999
CAS Number: 676348-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR