5-Methyl-4-(4-methylphenyl)thiophene-3-carbohydrazide
Supplier: Matrix Scientific
029846-500MG
101828-972EA
290.77
USD
101828-972
5-Methyl-4-(4-methylphenyl)thiophene-3-carbohydrazide
5-Methyl-4-(4-methylphenyl)thiophene-3-carbohydrazide
Hazard: Irritant
Formula:
C₁₃H₁₄N₂OS |
MDL Number:
MFCD03419885 CAS Number: 884497-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...