6-tert-Butyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carbohydrazide

Supplier: Matrix Scientific

029842-500MG
101828-964EA 290.77 USD
101828-964
6-tert-Butyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carbohydrazide
6-tert-Butyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carbohydrazide

Hazard: Irritant

Formula: C₁₃H₂₀N₂OS
MDL Number: MFCD03419866
CAS Number: 438225-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR