6-Methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carbohydrazide

Supplier: Matrix Scientific

029841-500MG
101828-962EA 290.77 USD
101828-962
6-Methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carbohydrazide
6-Methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carbohydrazide

Hazard: Irritant

Formula: C₁₀H₁₄N₂OS
MDL Number: MFCD03419860
CAS Number: 438211-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR