5-Isopropylthiophene-3-carbohydrazide

Supplier: Matrix Scientific

029840-500MG
101828-960EA 290.77 USD
101828-960
5-Isopropylthiophene-3-carbohydrazide
5-Isopropylthiophene-3-carbohydrazide

Hazard: Irritant

Formula: C₈H₁₂N₂OS
MDL Number: MFCD03419854
CAS Number: 438221-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR