5-Propylthiophene-3-carbohydrazide

Supplier: Matrix Scientific

029839-500MG
101828-958EA 290.77 USD
101828-958
5-Propylthiophene-3-carbohydrazide
5-Propylthiophene-3-carbohydrazide

Hazard: Irritant

Formula: C₈H₁₂N₂OS
MDL Number: MFCD03419853
CAS Number: 438215-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR