5-Ethylthiophene-3-carbohydrazide

Supplier: Matrix Scientific

029838-500MG
101828-956EA 290.77 USD
101828-956
5-Ethylthiophene-3-carbohydrazide
5-Ethylthiophene-3-carbohydrazide

Hazard: Irritant

Formula: C₇H₁₀N₂OS
MDL Number: MFCD03419848
CAS Number: 438225-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR